missing translation for 'onlineSavingsMsg'
Learn More
Methyl salicylate, 99%, Thermo Scientific Chemicals Product Code.: 10606052

Methyl salicylate, 99%, Thermo Scientific Chemicals

Product Code. 10606052
500 mL, Glass bottle
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10606052

Brand: Thermo Scientific Acros 220495000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -8°C to 7°C
Color Colorless to Yellow
Density 1.1825g/mL
Boiling Point 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2CH3
Refractive Index 1.5350 to 1.5380
Quantity 500 mL
Beilstein 10,7
Merck Index 15,6193
Specific Gravity 1.1825
Solubility Information Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C)
Formula Weight 152.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl salicylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468287 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number220490025,220490050,220495000,220490010
Lot NumberA0443029 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number220490025,220490050,220495000,220490010
Lot NumberA0457729 Certificate TypeCertificate of Analysis Date20/11/2023 Catalog Number220490025,220490050,220495000,220490010
Lot NumberA0377808 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number220490025,220490050,220495000,220490010
Lot NumberA0392685 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number220490025,220490050,220495000,220490010
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl salicylate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.