missing translation for 'onlineSavingsMsg'
Learn More
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™ Product Code.: 10517264

Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™

Product Code. 10517264
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10517264

Brand: Thermo Scientific Acros 426941000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

CAS 2206-27-1
Melting Point 20.2°C
Color Colorless
Density 1.1900g/mL
Boiling Point 190.0°C
Flash Point 88°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C2H6OS
MDL Number MFCD00002090
Quantity 100 mL
Specific Gravity 1.19
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
Solubility Information Solubility in water: soluble
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Molecular Weight (g/mol) 84.17
PubChem CID 75151
Viscosity 2.4 mPa.s (20°C)
Formula Weight 84.18
Grade NMR
Physical Form Liquid
Chemical Name or Material Methyl sulfoxide-d6, For NMR, 99.9 atom% D
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467742 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number426941000
Lot NumberA0465549 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number426941000
Lot NumberA0462714 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number426941000
Lot NumberA0461601 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number426941000
Lot NumberA0460018 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog Number426941000
Product Identifier
  • Methyl sulfoxide-d6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.