missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
pH | 7 |
Melting Point | -127°C |
Color | Colorless |
Boiling Point | 97°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 25mbar at 20°C |
Viscosity | 2.2 mPaS at 20°C |
Formula Weight | 60.1g/mol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
Safety and Handling



Product Identifier
- 1-Propanol
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Propan-1-ol, for HPLC, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction