missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Vinyl Ketone, Stabilized ca 95%, Thermo Scientific Chemicals
£32.76 - £354.10
Chemical Identifiers
CAS | 78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00008777 |
InChI Key | FUSUHKVFWTUUBE-UHFFFAOYSA-N |
Synonym | methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone |
PubChem CID | 6570 |
ChEBI | CHEBI:48058 |
IUPAC Name | but-3-en-2-one |
SMILES | CC(=O)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10552331
|
Thermo Scientific Acros
128000050 |
5 mL | Glass bottle |
£32.76
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16221612
|
Thermo Scientific Acros
128001000 |
100 mL | Glass bottle |
£104.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10418290
|
Thermo Scientific Acros
128000010 |
1 L | Glass bottle |
£217.08
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944821
|
Thermo Scientific Acros
128000025 |
2.5 L | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5 | |
70.09 | |
FUSUHKVFWTUUBE-UHFFFAOYSA-N | |
6570 | |
but-3-en-2-one |
C4H6O | |
MFCD00008777 | |
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone | |
CHEBI:48058 | |
CC(=O)C=C |
Specifications
78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5 | |
0.8640g/mL | |
−7°C | |
ca. 95% | |
H2C=CHCOCH3 | |
MFCD00008777 | |
0.864 | |
FUSUHKVFWTUUBE-UHFFFAOYSA-N | |
but-3-en-2-one | |
70.09 | |
CHEBI:48058 | |
ca. 95% | |
Methyl vinyl ketone |
Brown to Colorless | |
80.0°C | |
Authentic | |
C4H6O | |
1.4100 to 1.4120 | |
01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193 | |
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone | |
CC(=O)C=C | |
0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone | |
6570 | |
70.09 | |
Liquid |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methyl vinyl ketone, stabilized
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only