Learn More
Methylcobalamin hydrate, 97%, Thermo Scientific Chemicals
£59.50 - £238.00
Chemical Identifiers
CAS | 288315-09-3 |
---|---|
Molecular Formula | C63H91CoN13O14P- |
Molecular Weight (g/mol) | 1344.405 |
MDL Number | MFCD00149221 |
InChI Key | QXOMMGINPOYKPR-YHKGLSPZSA-L |
Synonym | methylcobalamin |
PubChem CID | 134128059 |
IUPAC Name | carbanide;cobalt(2+);[(2S,3R,4S,5R)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2S)-1-[3-[(1S,2S,3S,5Z,7R,10Z,12R,13R,15Z,17R,18R,19S)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-o |
SMILES | [CH3-].CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)([O-])OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[Co+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439113
|
Thermo Scientific Alfa Aesar
A11176.MC |
100 mg |
£59.50
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449113
|
Thermo Scientific Alfa Aesar
A11176.ME |
500 mg |
£238.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.
Solubility
Partially soluble in cold water, hot water.
Notes
Store at -20°C. Incompatible with strong oxidizing agents.
Chemical Identifiers
288315-09-3 | |
1344.405 | |
QXOMMGINPOYKPR-YHKGLSPZSA-L | |
134128059 | |
[CH3-].CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)([O-])OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[Co+2] |
C63H91CoN13O14P- | |
MFCD00149221 | |
methylcobalamin | |
carbanide;cobalt(2+);[(2S,3R,4S,5R)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2S)-1-[3-[(1S,2S,3S,5Z,7R,10Z,12R,13R,15Z,17R,18R,19S)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-o |
Specifications
288315-09-3 | |
MFCD00149221 | |
Partially soluble in cold water,hot water. | |
[CH3-].CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)([O-])OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[Co+2] | |
1344.405 | |
1344.41 (Anhydrous) | |
Methylcobalamin hydrate |
C63H91CoN13O14P- | |
methylcobalamin | |
QXOMMGINPOYKPR-YHKGLSPZSA-L | |
carbanide;cobalt(2+);[(2S,3R,4S,5R)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2S)-1-[3-[(1S,2S,3S,5Z,7R,10Z,12R,13R,15Z,17R,18R,19S)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-o | |
134128059 | |
98% |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350
missing translation for 'einecsNumber' : 236-535-3
missing translation for 'rtecsNumber' : GG3745000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only