missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, pure Product Code.: 10045700

Thermo Scientific Chemicals Methylene Blue hydrate, pure

Product Code. 10045700
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10045700

Brand: Thermo Scientific Chemicals 414251000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

Melting Point 100.0°C to 110.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Infrared Spectrum Authentic
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
Formula Weight 319.85
Loss on Drying 22% max.
Percent Purity ≥70% (TiCl3-titration)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methylene Blue hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469284 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number414251000,414250025,414255000
Lot NumberA0461903 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog Number414251000,414250025,414255000
Lot NumberA0450270 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog Number414251000,414250025,414255000
Lot NumberA0402362 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number414251000,414250025,414255000
Lot NumberA0413614 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number414251000,414250025,414255000
compliance-icons
Product Identifier
  • Methylene Blue hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Phenothiazin-5-ium,3,7-bis(dimethylamino)-, chloride, hydrate, Methylene Blue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methylene Blue hydrate, pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.