missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylthymol blue, sodium salt , indicator grade
£32.76 - £107.54
Chemical Identifiers
CAS | 1945-77-3 |
---|---|
Molecular Formula | C37H44N2Na4O13S |
Molecular Weight (g/mol) | 848.779 |
MDL Number | MFCD00148905 |
InChI Key | LGVVZVZPOQJZKT-UHFFFAOYSA-N |
PubChem CID | 131850582 |
IUPAC Name | 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10560744
|
Thermo Scientific Chemicals
394210010 |
1 g | Glass bottle |
£32.76
1g |
In Stock
Log in to see stock. |
||||
10085573
|
Thermo Scientific Chemicals
394210050 |
5 g | Glass bottle |
£107.54
5g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
1945-77-3 | |
848.779 | |
LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
C37H44N2Na4O13S | |
MFCD00148905 | |
131850582 | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
Specifications
1945-77-3 | |
MFCD00148905 | |
LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] | |
848.779 | |
844.76 | |
pH 6.5 (yellow) to pH 8.5 (blue):Pass Test | |
Crystalline Powder |
C37H44N2Na4O13S | |
Solubility in water: 460g/L (20°C). | |
Authentic | |
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium | |
131850582 | |
ACS Reagent | |
Red | |
Methylthymol blue, tetrasodium salt |
Safety and Handling
Product Identifier
- Methylthymol Blue, tetrasodium salt
RUO – Research Use Only