Learn More
Metro™ Double-Deep qwikTRAK™ Track Sets
Create tracks up to 21 ft.
£820.00 - £3295.00
Specifications
For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system |
---|---|
Product Type | Track Set |
Product Code | Brand | Depth (English) | Depth (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Depth (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
15907011
|
Metro™
LBTS8NA |
96 in. | 243.84 cm | 11 lb. | 5 kg |
£1395.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15917011
|
Metro™
LBTS9NA |
108 in. | 274.32 cm | 13 lb. | 5.9 kg |
£1515.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15987001
|
Metro™
LBTS6NA |
72 in. | 182.88 cm | 9 lb. | 4.1 kg |
£835.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15957001
|
Metro™
LBTS2.5NA |
30 in. | 76.2 cm | 4 lb. | 1.8 kg |
£820.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15956991
|
Metro™
LBTS10NA |
120 in. | 304.8 cm | 14 lb. | 6.4 kg |
£1635.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15997001
|
Metro™
LBTS7NA |
84 in. | 213.36 cm | 10 lb. | 4.5 kg |
£950.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15977001
|
Metro™
LBTS21NA |
252 in. | 640.08 cm | 29 lb. | 29 kg |
£3295.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15967001
|
Metro™
LBTS20NA |
240 in. | 609.6 cm | 27 lb. | 12.3 kg |
£3175.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15966991
|
Metro™
LBTS11NA |
132 in. | 335.28 cm | 15 lb. | 6.8 kg |
£1760.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15976991
|
Metro™
LBTS12NA |
144 in. | 365.76 cm | 17 lb. | 7.7 kg |
£1885.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15996991
|
Metro™
LBTS14NA |
168 in. | 426.72 cm | 19 lb. | 8.6 kg |
£2125.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15927001
|
Metro™
LBTS17NA |
204 in. | 518.16 cm | 23 lb. | 10.5 kg |
£2820.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15986991
|
Metro™
LBTS13NA |
156 in. | 396.24 cm | 18 lb. | 8.2 kg |
£2005.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15907001
|
Metro™
LBTS15NA |
180 in. | 457.2 cm | 21 lb. | 9.5 kg |
£2570.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15937001
|
Metro™
LBTS18NA |
216 in. | 548.64 cm | 25 lb. | 11.4 kg |
£2940.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15917001
|
Metro™
LBTS16NA |
192 in. | 487.68 cm | 22 lb. | 10 kg |
£2690.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15947001
|
Metro™
LBTS19NA |
228 in. | 579.12 cm | 26 lb. | 12 kg |
£3055.00
Each |
Estimated Shipment: 02-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
- Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
- 17-823-756 (LBTS2.5NA) includes a joining kit to connect the track to other track sets
Specifications
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system | |
Track Set |