missing translation for 'onlineSavingsMsg'
Learn More

Metro™ HD Super Stainless Laboratory Worktable Accessory, Utility Racks

For use with HD Super worktables

Brand:  Metro™ 654SRC

 View more versions of this product

Product Code. 15986741

  • £139.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Choice of racks for use with tables with overheads

Specifications

Specifications

Utility Rack
11 cm
Overhead 60 in. Worktable Models
4.9 cm
131.8 cm
4.37 in.
Chrome-plated
1.93 in.
51.96 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers