Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Black Phenolic Top with 3-Sided Frame
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Brand:  Metro™ LT48XUPB3
 View more versions of this product
Product Code. 15977041
  • £1445.00 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These worktables provide the ideal product platform, are modular by design and offer ultimate flexibility based on the application requirements.
  • Advanced polymer construction for corrosive lab environments
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
Stationary Table
Ideal for lab
101.6 cm
47.75 in.
600 lb.
30 in.
Polymer
47.75 x 30 x 40.5 in.
40.5 in.
3 Sided frame
121.3 cm
273 kg
76.2 cm