missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Solid and Wire Shelves

Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Solid and Wire Shelves make a smart addition to any workplace.

Brand:  Metro™ MW401

Additional Details : Weight : 19.07200kg

 View more versions of this product

Product Code. 15967171

  • £635.00 / Each
Estimated Shipment: 19-11-2024
to see stock.
Add to basket

Description

Description

Stainless steel, chrome shelf and handle material.

Cart

  • Load capacity: 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta™ shelves
  • Center shelf can be positioned at any height in 1 in. (2.5cm) increments
  • One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability
  • 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in. (53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
  • Three tiers
  • 375 lbs (170kg) weight capacity per cart
Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting
Wire Shelves
  • Chrome-plated or stainless-steel shelves available with trademarked truss design for added strength
  • Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Specifications

Utility Cart
3-tier Model
Transporting materials that require an expanse of flat, solid surface
38 in.
Super Erecta™ 2 Chrome plated wire shelves and one Stainless Steel solid (flat) shelf with Plastic split sleeves, 2 one-piece Chrome plated handles, 4 in. dia. caster, 3.5 in. donut bumpers
61 cm
170 kg
Stainless Steel, Chrome
43 lb.
46 cm
NSF
24 x 18 x 38 in. (61 x 46 x 97 cm)
Chrome
97 cm
24 in.
375 lb.
3
19.5 kg
18 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers