missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MW Series Utility Cart with Solid and Wire Shelves
Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Solid and Wire Shelves make a smart addition to any workplace.
£635.00 - £750.00
Specifications
Product Type | Utility Cart |
---|---|
Certifications/Compliance | NSF |
Description | 3-tier Model |
For Use With (Application) | Transporting materials that require an expanse of flat, solid surface |
Handle Material | Chrome |
Product Code | Brand | Dimensions (L x W x H) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Includes | Price | Quantity & Availability | ||||
15977171
|
Metro™
MW402 |
18 x 30 x 39 in. (45.72 x 76.2 x 99 cm) | Super Erecta™ 2 Chrome plated wire shelves and one Stainless Steel solid (flat) shelf with Plastic split sleeves, 2 one-piece Chrome plated handles, 4 in. dia. caster, 3.5 in. donut bumpers |
£690.00
Each |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
15967171
|
Metro™
MW401 |
24 x 18 x 38 in. (61 x 46 x 97 cm) | Super Erecta™ 2 Chrome plated wire shelves and one Stainless Steel solid (flat) shelf with Plastic split sleeves, 2 one-piece Chrome plated handles, 4 in. dia. caster, 3.5 in. donut bumpers |
£635.00
Each |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
15987171
|
Metro™
MW404 |
36 x 21 x 38 in. (91 x 53 x 97 cm) | Super Erecta™ 2 Chrome plated wire shelves and one Stainless Steel solid (flat) shelf with Plastic split sleeves, 2 one-piece Chrome plated handles, 5 in. dia. caster, 3.5 in. donut bumpers |
£750.00
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||||
Description
Stainless steel, chrome shelf and handle material.
Cart
- Load capacity: 375 lb. (170.1kg)
- Fully interchangeable Super Erecta™ shelves
- Center shelf can be positioned at any height in 1 in. (2.5cm) increments
- One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability
- 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in. (53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
- Three tiers
- 375 lbs (170kg) weight capacity per cart
- 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
- Raised edges help contain spills and make cleanup easier
- Sturdy corner castings facilitate mounting
- Chrome-plated or stainless-steel shelves available with trademarked truss design for added strength
- Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Utility Cart | |
3-tier Model | |
Chrome | |
170 kg | |
Stainless Steel, Chrome |
NSF | |
Transporting materials that require an expanse of flat, solid surface | |
375 lb. | |
3 |