missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and Solid HD Shelf
Lab worktable provides safety in the laboratory with corrosion-proof performance and stainless steel construction. Features a solid HD shelf and a stainless top/backsplash .
Brand: Metro™ LTS48PB
This item is not returnable.
View return policy
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Stationary Table | |
47.75 x 30 x 36 in. | |
36 in. | |
Solid HD Shelf | |
121.2 cm | |
273 kg | |
76.2 cm |
Silver | |
Laboratory | |
91.4 cm | |
47.75 in. | |
600 lb. | |
30 in. | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction