Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
High-density, easy access storage
Brand:  Metro™ TTA24S
 View more versions of this product
Product Code. 15997281
  • £876.00 / Each
Estimated Shipment: 27-08-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts
Order Info
Shelves are ordered separately
Specifications
Specifications
Stationary Intermediate Unit Kit
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
2185 mm
8.5 lb.
24 in.
Stainless Steel
None
86 in.
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
3.8 kg
61 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits > Stainless Steel; Shelf width: 610mm (24 in.)