Learn More
Metro™ MetroMax™ iQ Polymer Shelving Accessory, Shelf Ledge, Stackable
2 in. ledges provide a secure lip to secure items during transport or to meet seismic or other safety requirements
Brand:  Metro™ MXLS18-2S
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15997211
  • £28.00 / Each
Estimated Shipment: 25-09-2024
to see stock.
Add to basket
Description
Description
Easily attach with no tools to MetroMax i solid or open grid shelves. Other sizes and styles available.
  • Snap on and off quickly
  • Easily stacked to contain larger items
  • Corrosion proof
  • Ships assembled
Specifications
Specifications
18 in.
2 in.
45.7 cm
5 cm
Metro™ MetroMax i™ and Q™ shelving
Stainless Steel