Learn More
Thermo Scientific Chemicals Metronidazole, 99%
Metronidazole, CAS# 443-48-1, is a member of the class of imidazoles with limited antibiotic activity that acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria.
£39.80 - £998.97
Chemical Identifiers
CAS | 443-48-1 |
---|---|
Molecular Formula | C6H9N3O3 |
Molecular Weight (g/mol) | 171.16 |
MDL Number | MFCD00009750 |
InChI Key | VAOCPAMSLUNLGC-UHFFFAOYSA-N |
Synonym | metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol |
PubChem CID | 4173 |
ChEBI | CHEBI:6909 |
IUPAC Name | 2-(2-methyl-5-nitroimidazol-1-yl)ethanol |
SMILES | CC1=NC=C(N1CCO)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10691063
|
Thermo Scientific Chemicals
210340050 |
5 g | Glass bottle |
£39.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666792
|
Thermo Scientific Chemicals
210341000 |
100 g | Plastic bottle |
£296.48
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10678124
|
Thermo Scientific Chemicals
210345000 |
500 g | Plastic bottle |
£998.97
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Metronidazole is a synthetic nitroimidazole derivative with antiprotozoal and antibacterial activities and acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria
• Metronidazole is considered a prodrug that produces nitroso-containing intermediates in the cell that can covalently bind DNA, disrupting helical structure, inducing DNA strand breaks, and inhibiting bacterial nucleic acid synthesis, that results in bacterial cell death
Applications
• Metronidazole shows anti-parasitic and anti-amoebic activities.
• It has been used to prevent the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such as Peptostreptococcus and Clostridia. This compound affects a high percentage of gram-negative bacteria and has higher tissue penetration that gives it an advantage compared to other compounds.
• This compound was used initially to suppress protozoans such as Entamoeba histolytica, Giardia lamblia, and Trichomonas vaginalis.
• It has been commonly applied for the eradication of Helicobacter pylori.
• It also has a radio-sensitizing effect on hypoxic tumor cells and can have a mutagenic activity in in vivo animal models.
Chemical Identifiers
443-48-1 | |
171.16 | |
VAOCPAMSLUNLGC-UHFFFAOYSA-N | |
4173 | |
2-(2-methyl-5-nitroimidazol-1-yl)ethanol |
C6H9N3O3 | |
MFCD00009750 | |
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol | |
CHEBI:6909 | |
CC1=NC=C(N1CCO)[N+](=O)[O-] |
Specifications
443-48-1 | |
100.0 | |
White to Yellow | |
99% | |
MFCD00009750 | |
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol | |
VAOCPAMSLUNLGC-UHFFFAOYSA-N | |
2-(2-methyl-5-nitroimidazol-1-yl)ethanol | |
4173 | |
171.16 | |
Crystalline Powder |
98.5 | |
159°C to 163°C | |
Authentic | |
C6H9N3O3 | |
15, 6236 | |
Solubility in water: 1g/100mL (20°C). Other solubilities: soluble in diluted acids,0.5g/100mL ethanol- <0.05g/100mL ether (20°C),<0.05g/100mL chloroform (20°C),sparingly soluble in dmf | |
CC1=NC=C(N1CCO)[N+](=O)[O-] | |
171.16 | |
CHEBI:6909 | |
99% | |
Metronidazole |
Safety and Handling
GHS H Statement
May cause cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-136-1
- Metronidazole
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
Hazard Statement
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only