missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Redox Buffer Solutions

Storage and refill solutions for redox electrodes.

Brand:  Mettler Toledo™ 51350060

37.79 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 10494071

  • £40.60 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Includes a quality inspection certificate which guarantees the stated values and traceability.

Certifications

NIST traceable

TRUSTED_SUSTAINABILITY
Specifications

Specifications

∼100°C
Buffer Solution
Quality inspection certificate
250 mL Bottle
Completely Miscible
Clear, Yellow
Redox Buffer; UH = 427mV
220 mV
7
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    22

    Hours

    4

    Minutes

    36

    Seconds

    34
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Redox Buffer Solutions > UH = 427mV; Packaging: 250mL bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.