Learn More
METTLER TOLEDO™ S479 SevenExcellence™ pH/Conductivity/DO Benchtop Meter Kit
The S479 meter kit gives you everything you need to start measuring pH, conductivity, dissolved oxygen, BOD, resistivity, salinity, TDS, Redox/ORP, and ISFET all on one meter. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.
Brand: METTLER TOLEDO™ 30092988
5970.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: SevenExcellence pH/mV/Conductivity/DO triple channel meter (instrument with a pH/mV, conductivity and DO expansion unit); as well as an InLab™ ExpertPro-ISM (pH), InLab 731-ISM (conductivity) and InLab OptiOx (digital DO) sensor; pH buffers; conductivity standards; and zero-oxygen calibration tablets; guides to pH, conductivity, and DO measurement; uPlace™ electrode holder, semi-transparent cover, operating instructions, installation, LabX™ direct pH PC software, declaration of conformity and test certificate
Description
S479 meter kit containing SevenExcellence meter, electrode, and accessories to perform pH, conductivity, dissolved oxygen, resistivity, salinity, TDS, BOD, Redox/ORP, and ISFET measurements
- The SevenExcellence can be expanded with additional modules to measure pH, conductivity, resistivity, ISE, salinity, TDS, Redox/ORP, and ISFET
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
- Preventative maintenance and service available through METTLER TOLEDO Service
Specifications
SevenExcellence pH/Cond/DO meter S479-K | |
S479 pH/Conductivity/DO meter, InLab Expert Pro-ISM pH electrode (1.2 m cable), InLab 731-ISM conductivity electrode, InLab OptiOx Optical dissolved oxygen electrode (1.8 m cable), InLab OptiOx calibration tube, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, 2 buffer sachets each for pH 1413 mS/cm and 12.88 mS/cm), EasyDirect pH activation key, test certificate, pH measurement guide, conductivity/DO measurement guide | |
250 analyses (20.000 data points) | |
S479 SevenExcellence pH/Conductivity/DO Meter Kit |
Multi-channel | |
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
S479 pH/Conductivity/DO Kit |
Special Offers
- Multiple Manufacturers