missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ SevenExcellence™ S470 pH/Conductivity Benchtop Meter

Product Code. p-4519547 Shop All Mettler Toledo Products
Click to view available options
Model:
Meter, Standard Kit
Meter, USP/EP Compliance Kit
S470 pH/Conductivity Meter
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13496618

Brand: METTLER TOLEDO™ undefined

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The S470 meter measures pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET. It can be expanded with additional modules to measure dissolved oxygen, BOD, and ISE. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.

  • pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET meter than can be expanded with additional measurement modules to measure dissolved oxygen, ISE, and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
Note: pH and conductivity electrodes are not included; sold separately.
TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
Accuracy (Conductivity) ±0.5%
Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Calibration Points (Conductivity) 13 Pre defined and 20 user defined standard
Calibration Points (ISE) Max. 5 points, 8 pre defined and 20 user defined buffer group
Calibration Points (pH) Max. 5 points, 8 pre defined and 20 user defined buffer group
Data Points 20,000
Display Type TFT
Range (Conductivity) 0.001 μS/cm to 2000 mS/cm
Range (mV) -2000 to 2000 mV
Range (pH) -2 to 20
Range (Resistivity) 0.01 to 100 Mohm/cm
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.001 mg/L to 1000 g/L
Resolution (ISE) 0.001 to 1
Resolution (pH) 0.001/0.01/0.1
Temperature Range (Metric) -30°C to +130°C
Includes S470 pH/conductivity meter, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, 2 standard sachets each for 1413 uS/cm and 12.88 mS/cm, EasyDirect pH activation key, test certificate
Model S470 pH/Conductivity Meter
Range (ISE) 0 to 999999mg/L
Resolution (Conductivity) 0.001μS/cm to 2000mS/cm
Resolution (mV) 0.1/1mV
Resolution (Resistivity) 0.001 to 1
Resolution (Salinity) 0.001 to 1
Resolution (TDS) 0.001 to 1
Temperature Accuracy ±0.1°C
Temperature Resolution 0.1°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.