missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.45 μm

Mixed cellulose esters (MCE) membrane, hydrophilic

Brand:  Merck HAWP14250

400.11 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 71

Additional Details : CAS Number : 9004-70-0 Weight : 0.05000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10400781

  • £441.00 / Pack of 50
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Designed for a wide range of analytical and research applications. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.45μm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications

Specifications

Not Autoclavable
White
0.45 μm
60 mL/min x cm2
Hydrophilic
50Pack
150 μm
Hydrophilic
142 mm
≥1.8 bar, air with water
MCE
Non-sterile
4 L/min/ cm2
Plain
50/Pk.
Filter Membranes
142 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MCE .45UM WH PL 142MM 50/PK

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers