missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.45 μm

Mixed cellulose esters (MCE) membrane, hydrophilic

Brand:  Merck HAWP03700

Code : 72

Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10537492

  • £199.00 / Pack of 100
Estimated Shipment: 09-09-2024
to see stock.

Description

Description

Designed for a wide range of analytical and research applications. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.45μm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications

Specifications

4L/min./cm2
≥1.8 bar, air with water
Millipore Aerosol Analysis Monitor
0.45 μm
HAWP
Clarification of aqueous solutions, particle analysis and microbiology analysis
HAWP
Thin absorbent pads
100Pack
Plain
75°C
Filter Membranes
1.4 in.
37 mm
Not Autoclavable
White
MCE
Non-sterile
60 mL/min x cm2
4 L/min/ cm2
Hydrophilic
Plain
150μg/cm2
167°F
150 μm
Hydrophilic
3.7 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • monitor refills MCE 0.45µm WH PL 37mm

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers