missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free
Composed of mixed cellulose esters (nitrate and acetate)
Brand: Merck GSTF01300
Code : 71
Additional Details : Weight : 0.10000kg
Description
Composed of mixed cellulose esters (nitrate and acetate)
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.Features & Benefits: - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes, colored black or white, with or without a gridded surface - Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methodsSpecifications
Not Autoclavable | |
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free | |
Non-sterile | |
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.). | |
Filter Membranes | |
13 mm |
White | |
0.22 μm | |
18 mL/min x cm2 | |
Plain | |
Hydrophilic | |
13 mm |