missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free
Composed of mixed cellulose esters (nitrate and acetate)
£111.00 - £447.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Material | Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free |
Sterility | Non-sterile |
For Use With (Application) | Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.). |
Product Code | Brand | Diameter (Metric) Filter | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Pore Size | Price | Quantity & Availability | ||||
10319750
|
Merck
HATF02500 |
25 mm | 0.45 μm |
£117.00
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10247820
|
Merck
HATF14250 |
142 mm | 0.45 μm |
£447.00
Pack of 50 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
10309610
|
Merck
GSTF02500 |
25 mm | 0.22 μm |
£129.00
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10004690
|
Merck
HATF09025 |
90 mm | 0.45 μm |
£155.00
Pack of 25 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
10126490
|
Merck
GSTF01300 |
13 mm | 0.22 μm |
£118.00
Pack of 100 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
10607321
|
Merck
HATF04700 |
47 mm | 0.45 μm |
£196.00
Pack of 100 |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10480211
|
Merck
HATF01300 |
13 mm | 0.45 μm |
£111.00
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11596395
|
Merck
GSTF04700 |
47 mm | 0.22 μm |
£186.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Composed of mixed cellulose esters (nitrate and acetate)
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.Features & Benefits: - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes, colored black or white, with or without a gridded surface - Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methodsSpecifications
Not Autoclavable | |
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free | |
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.). | |
Hydrophilic |
White | |
Non-sterile | |
Plain |