missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free

MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free composed of mixed cellulose esters (nitrate and acetate).

Brand:  Merck GSTF04700


 View more versions of this product

Product Code. 11596395

  • £199.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Composed of mixed cellulose esters (nitrate and acetate)

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features and Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Reusable
Disc
Cellulose
Non-sterile
Clarification of fluids, Sterilizing filtration of tissue culture media, additives, antibiotics, Air monitoring, Particulate analysis, Particulate removal, Biological assays (receptor binding studies, TCA precipitates, etc.).
Filter Membranes
1.9 in.
47 mm
White
Membrane Filter
MCE
0.22 μm
18 mL/min. x cm2
Plain
Hydrophilic
47 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MCE TRITON FREE .22UM WH PL 47MM 100/PK

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers