Learn More
MFN1 Mouse anti-Human, CoraLite 488, Clone: 3F11C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66776-100UL
Description
The MFN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MFN1. This antibody recognizes human antigen. The MFN1 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Specifications
MFN1 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence, Flow Cytometry | |
3F11C11 | |
CoraLite 488 | |
100 μL | |
55669 | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed