missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Micro Double Spatula, Nickel
Double-ended for versatility and micro-sized for easy reach into small vials. Bochem™ Micro Double Spatula, Nickel has a flat end, and a rounded end, and is constructed of resistant Nickel (99.5%).
Brand: Bochem™ 3047
This item is not returnable.
View return policy
Description
- Double-ended.
- Micro-sized.
- Round end and flat end.
- Nickel (99.5%) construction.
Specifications
Double Ended Micro Spatula | |
8 mm | |
130 mm |
40 mm | |
Nickel (99.5%) |