Learn More
3M™ Microbial Luminescence System MLSII
A rapid microbial detection system using ATP bioluminescence in a microwell based System to detect the presence of microbial ATP in Ultra Heat Treated (UTH) and Extended Shelf Life (ESL) dairy and dairy end products.
Brand: 3M™ 7000032756
This item is not returnable.
View return policy
Description
- Microplate, 5 injector-dispenser luminometer designed for microbial detection.
- Results available approximately 48 hours sooner than traditional methods.
- Reduces stock holding requirements and improves responsiveness to customers.
- Time saved improves cash flow.
- Allows quality performance to be tracked over time.
Specifications
1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Microbial Luminescence System MLSII
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu