missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff

Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
Description
- Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
- Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
- Made with a soft, flexible nitrile formulation
- Fully textured for a reliable, consistent grip
- For use with:
- Administering drugs
- Emergency services
- Equipment repair and maintenance
- Extra protection over wrist and forearm
- Inspection, selecting, checking parts
- Laboratory analysis and testing
- Paint and spray shops
- Patient examinations
- Raw material sample collection
- Tire rotation and change
Greener Choice Claims
- Environmental benefits include:
- Recyclable

Specifications
Specifications
Certifications/Compliance | NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600 |
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 455 1-4 |
Applications | Healthcare, Laboratory |
Color | Black |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 6.3 mil |
Finger Thickness (Metric) | 0.16 mm |
Latex Free | Yes |
Material | Nitrile |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Special Offers
-
Multiple Manufacturers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff >
Spot an opportunity for improvement?Share a Content Correction