missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Microflex™ Variable-Speed Pump System

Microflex™ Variable-Speed Pump Systems are easy-to-use low-flow pumps that feature reversible pumping, remote start and stop, and rapid tubing changes. Ideal for consistent fluid delivery in analyzer and other low-flow laboratory pumping applications.

Brand:  Ismatec™ MFLX77122-30

1121.00 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 17477563

  • £1320.00 / 1 unit

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
  • Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings
  • Separate single-turn speed control and clockwise/off/counter-clockwise switch with green LED power indicator, thus maintaining speed setting when you turn pump on/off or reverse direction
  • Reversible permanent magnet DC motor allows fluid purging before or after pumping
  • Remote control connections on back of pump includes start/stop pump with a contact closure
  • Quick-loading Microflex™ pump head has average fixed occlusion for tubing, no occlusion adjustments required
Specifications

Specifications

1
Analog
0.002 to 1.65 mL/min.
Yes
4
50/60 Hz
Microflex™ Compact Single-Channel Pump
Analog
1.7 to 10 rpm
1
Microbore 0.19, 0.25, 0.51, 0.89, 1.14, 1.42, 2.06, 2.79 mm ID
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.