missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with Supor™ Membrane

Ready-to-use, sterile and individually packaged MicroFunnel filter funnels reduce chances of cross contamination.

Brand:  Cytiva (formerly Pall Lab) 4803

Additional Details : Weight : 2.00450kg

 View more versions of this product

Product Code. 17184401

  • £550.00 / Pack of 50
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • Easy squeeze removal of funnel to access membrane – no twisting means less RSI and membrane tearing
  • Proven membrane performance
  • For research use only
Specifications

Specifications

100 mL
50/Pk.
Gridded Filtering Funnel
0.2 μm
Sterile
Supor (PES)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers