Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ ST Filters Funnel with GN-6 Membrane
Microfunnel ST filter funnels are available in gamma-irradiated, double-bagged packaging saving valuable time during entry into cleanrooms and hoods.
Brand:  Cytiva (formerly Pall Lab) 4811
Additional Details : Weight : 1.86200kg
 View more versions of this product
Product Code. 17164411
  • £446.00 / Pack of 40
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • For research use only
Specifications
Specifications
100 mL
Sterile
GN-6 Metricel Mixed Cellulose Ester
0.45 μm
Filtering Funnel