Learn More
Ansell Microgard™ 2500 Standard Overboots with ESD PVC Soles Product Code.: 15536554
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Ansell Microgard™ 2500 Standard Overboots with ESD PVC Soles
Product Code. 15536554 Shop All Ansell Products
Product Code. 15536554
Brand: Ansell WH25B0040700
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Add to basket
Offers exceptional mechanical strength, liquid and particulate protection. Ansell Edmont™ Microgard™ 2500 Standard Overboots with ESD PVC Soles provide foot and calf protection from blood-borne pathogens and infective agents.
  • Microporous polypropylene laminate
  • Fits Euro sizes 42 to 46 (UK 8 to 11)
  • Elasticated opening
  • ESD PVC anti-slip sole
  • Adjustable Shoe Tie
  • Bound seams
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Color White
Closure Type Elastic with Adjustable Ties
Sole Type ESD PVC
Model WH25-B-00-407-00
Seam Type Bound
Packaging Quantity 150 Pairs
Product Type Boot Covers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell Microgard™ 2500 Standard Overboots with ESD PVC Soles >