missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 260
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filters, Grade 260 are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
£23.60 - £92.70
Specifications
Basis Weight | 143 g/m2 |
---|---|
Filter Type | Microfilter |
Format | Disc |
Material | Borosilicate Glass |
Pore Size | 1 μm |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11714103
|
Fisherbrand™
|
47 mm |
£57.25
Pack of 50 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
11744616
|
Fisherbrand™
|
54.8 mm |
£27.60
Pack of 50 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
11754616
|
Fisherbrand™
|
60 mm |
£29.10
Pack of 50 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
11704103
|
Fisherbrand™
|
25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11734616
|
Fisherbrand™
|
40.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11784556
|
Fisherbrand™
|
100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11744103
|
Fisherbrand™
|
124 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11734103
|
Fisherbrand™
|
109 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11764616
|
Fisherbrand™
|
69.8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11724103
|
Fisherbrand™
|
90 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Contains no binder
- Suitable for use at and up to 500°C
- Not affected by humidity
- Suitable for micro-filtration due to fine fibers
- High-loading capacity
- High chemical purity
- made with demineralized water
- High permeability to air
Applications:
- Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
- Sediments content in petroleum products
- Ligand binding test
- Air pollution monitoring
- Membranes pre-filters
- Radio-immuno tests
- Suspended solids in water, smokes
- Liquid scintillation counting
- Cells collection
- Grease content in dairy products
Specifications
143 g/m2 | |
Disc | |
1 μm | |
Membrane pre-filters, radio-immuno tests, liquid scintillation counting | |
Non-hygroscopic | |
0.027 in. | |
Filter |
Microfilter | |
Borosilicate Glass | |
200 sec. | |
260 | |
500°C | |
0.7 mm |