missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microglass Fiber Filters, Grade 261

Made with 100% borosilicate micro-glass fibers and are binder free. Fisherbrand™ Microglass Fiber Filters are particularly suitable for micro-filtration because of very fine fibers.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.03240kg

 View more versions of this product

Product Code. 11704083

  • £65.48 / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • High loading capacity
  • Excellent levels of particles retention (pore size 0.7 to 2.7μm)
  • Chemical resistance to many solvents and reagents
  • Non-hygroscopic and biologically inert
  • HIgh permeability against the passing of air
  • Suitable for use at and up to 500°C
Applications: filtration of industrial water, ligand binding test, proteins filtration, radio-immuno tests, suspended sollids in water (EN872 norm), drinking water filtration, cells collection
Specifications

Specifications

52 g/m2
Disc
1.2 μm
Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection
Non-hygroscopic
0.01 in.
Filter
Microfilter
Borosilicate Glass
100 sec.
261
500°C
0.26 mm
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers