missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 261
Made with 100% borosilicate micro-glass fibers and are binder free. Fisherbrand™ Microglass Fiber Filters are particularly suitable for micro-filtration because of very fine fibers.
£20.25 - £245.00
Specifications
Basis Weight | 52 g/m2 |
---|---|
Format | Disc |
Pore Size | 1.2 μm |
Filtration Time | 100 sec. |
For Use With (Application) | Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection |
Product Code | Brand | Diameter (Metric) | Material | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Material | Format | Price | Quantity & Availability | |||
11512083
|
Fisherbrand™
|
25 mm | Borosilicate Glass | Disc |
£65.25
Pack of 100 |
In Stock
Log in to see stock. |
|||
11704083
|
Fisherbrand™
|
47 mm | Borosilicate Glass | Disc |
£65.48
Pack of 100 |
In Stock
Log in to see stock. |
|||
11532083
|
Fisherbrand™
|
70 mm | Borosilicate Glass | Disc |
£124.87
Pack of 100 |
In Stock
Log in to see stock. |
|||
11714083
|
Fisherbrand™
|
90 mm | Borosilicate Glass | Disc |
£196.08
Pack of 100 |
In Stock
Log in to see stock. |
|||
11724083
|
Fisherbrand™
|
110 mm | Borosilicate Glass | Disc |
£200.21
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
11734083
|
Fisherbrand™
|
125 mm | Borosilicate Glass | Disc |
£163.00
Pack of 100 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
11744083
|
Fisherbrand™
|
150 mm | Borosilicate Glass | Disc |
£245.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
11735255
|
Fisherbrand™
|
55 mm | Borosilicate Glass Microfiber | Disc |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11881733
|
Fisherbrand™
|
28 mm | Borosilicate Glass | Disc |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- High loading capacity
- Excellent levels of particles retention (pore size 0.7 to 2.7μm)
- Chemical resistance to many solvents and reagents
- Non-hygroscopic and biologically inert
- HIgh permeability against the passing of air
- Suitable for use at and up to 500°C
Specifications
52 g/m2 | |
1.2 μm | |
Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection | |
500°C |
Disc | |
100 sec. | |
261 | |
0.26 mm |