missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 261
Made with 100% borosilicate micro-glass fibers and are binder free. Fisherbrand™ Microglass Fiber Filters are particularly suitable for micro-filtration because of very fine fibers.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.80000kg
Description
- High loading capacity
- Excellent levels of particles retention (pore size 0.7 to 2.7μm)
- Chemical resistance to many solvents and reagents
- Non-hygroscopic and biologically inert
- HIgh permeability against the passing of air
- Suitable for use at and up to 500°C
Specifications
52 g/m2 | |
Disc | |
1.2 μm | |
Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection | |
Non-hygroscopic | |
0.01 in. | |
Filter |
Microfilter | |
Borosilicate Glass | |
100 sec. | |
261 | |
500°C | |
0.26 mm | |
28 mm |