missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Isolator Tool and Irradiated Covers

The irradiated angled head adapter with a 360° swivel works with a range of irradiated mop covers to efficiently clean and disinfect isolators, hoods, RABS and small equipment in clean and aseptic environments.

Brand:  Micronova™ QPIT-09

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13137666

  • £63.00 / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Description

Description

Help to simplify cleaning in compliance with USP 797

The Micronova Isolator Tool is available with a choice of stainless steel or plastic plates and the mop covers are available in a range of fabrics with optional foam layer to hold disinfectant and facilitate wringing. Designed for use with the SSU Series Universal Handles or STA Series threaded handles to fit through small passes and allow extended reach.

  • Isolator tool with choice of covers
  • Ergonomic cleaning for isolators, hoods, RABS and other small equipment
  • Modular and extendable handles fit through small passes and expand reach
  • 360° swivel, angled head adapter for easy application of cleaning agent
  • The stainless steel adapter (QDIT) is designed for use with SSU series QD handles
  • The plastic adapter (QPIT) is designed for use with ACME threaded handles
  • Covers available in a variety of fabrics for specialized cleaning
    • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
    • PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
    • Microfiber — 80% polyester, 20% polyamide, highly absorbent and effective wet or dry
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Specifications

Specifications

22.9 x 5.1 cm (9 x 2 in.)
360 Swivel
9 in.
22.9 cm
4.8 cm
Chemicals
1 Ea.
Isolators, Containment and Bio-hoods in Cleanroom and Buffer Areas
Acetal
Plastic Isolator Tool, ACME Threaded Handle Attachment
Black
Non-sterile
Not Autoclavable
1.9 in.
No
Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
Cleaning Inside BSC, Hoods and Other Small Equiment.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers