missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ NovaWipe™ Wipers
For semiconductor and microelectronics cleaning where the fast-drying, absorbent properties of PVA are key
Brand: Micronova™ WP1-79
This item is not returnable.
View return policy
Description
- With a 600% absorbency, they are soft and pliable for both wet and dry applications
- For general wiping, lapping and polishing applications
- Can be laundered and re-used
Specifications
NovaWipe™ Cleanroom Wipes | |
22.9 cm | |
17.8 cm | |
Reusable | |
Ideal for general wiping, lapping and polishing applications | |
10 Wipers/Pk. |
9 in. | |
7 in. | |
Polyvinyl Alcohol | |
9 x 7 in. (22.9 x 17.8 cm) | |
Workstations, laminar flow benches, walls, windows | |
0.0625 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction