Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Brand: Thermo Scientific™ N03394
2626.23 GBP valid until 2024-12-31
Use promo code "21998" to get your promotional price.
Description
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation.Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
Specifications
Spectrophotometric Verification Plate | |
Verification Plate | |
NPL (UK National Physical Laboratory) and NIST (US National Institute for Standards and Technology) traceable Spectrophotometric Verification Plate Certificate of Calibration Adapter for Spectrophotometric Verification Plate USB memory device including SkanIt™ software sessions and reference files Carrying case for Spectrophotometric Verification Plate |
Varioskan LUX Microplate Reader, Multiskan Sky, Multiskan Skyhigh | |
Varioskan™ |