Learn More
Thermo Scientific™ Aluminum Support, tube/plate rack, 96-well
Thermo Scientific™ aluminum support rack can be used for sample preparation, heat-sealing and automatic dispensing.
Brand: Thermo Scientific™ PRK1963
116.38 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Perform sample preparation, heat-sealing and automatic dispensing using Thermo Scientific™ aluminum support rack.
Available in 96- and 384-well format.
- For heat-sealing and automated dispensing
- Industry-standard footprint fits in robotic workstations
- Can be heated or cooled to assist with sample preparation
Includes:
Recommended for:
Sample handling, sample preparation, heat-sealing, automated dispensing
Specifications
Microplate Rack | |
Microplates (-well) | |
Rectangular | |
1 rack |
Blue | |
Aluminum | |
Real-Time PCR Systems or Thermal Cyclers |
Product Suggestions
Customers who viewed this item also viewed