missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Microsep™ Advance Centrifugal Devices with Omega Membrane

Microsep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 5 mL of biological sample.

Brand:  Cytiva (formerly Pall Lab) MCP030C46

646.00 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.



Additional Details : Weight : 1.06400kg

 View more versions of this product

Product Code. 17134751

  • £680.00 / Pack of 100
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
Specifications

Specifications

Red
PES
Non-sterile
0.5 to 5 mL (Sample)
EFA: 3.3 cm2
30 kDa
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers