Learn More
Cytiva (formerly Pall Lab) Microsep™ Advance Centrifugal Devices with Omega Membrane
Microsep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 5 mL of biological sample.
Brand: Cytiva (formerly Pall Lab) MCP001C46
711.55 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.
Additional Details : Weight : 1.54000kg
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
Yellow | |
PES | |
Non-sterile | |
0.5 to 5 mL (Sample) |
EFA: 3.3 cm2 | |
1 kDa | |
100/Pk. |