missing translation for 'onlineSavingsMsg'
Learn More

KARTELL Microtitration Plate

The Microtitration Plate has 96 sample wells, available in two shapes: flat bottom and V-bottom.

Brand:  KARTELL 02622/S

46.03 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.



Code : NEW

Additional Details : Weight : 2.18000kg

 View more versions of this product

Product Code. 12850923

  • £48.45 / Pack of 50
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

  • Multiple-well plates with 96 sample wells in clear PS.
  • Flat bottom or V-bottom wells, single wrapped sterile.
Specifications

Specifications

No Barcode
8 x 12
None
Polystyrene
96
Non-treated
Sterile
Standard
Not Autoclavable
Clear
0.281 mL
Sterile
Conical Bottom (V-well)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers