missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Luria-Bertani Agar, pre buffered capsules
Used for culturing E. coli.
Brand: Fisher BioReagents™ 12861670
Code : 32
Additional Details : Weight : 0.62980kg
Description
- Pre-buffered
Specifications
Room Temperature | |
Agar | |
25 x 20 g |
Capsules | |
25 x 20g |