missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Luria-Bertani Agar, pre buffered capsules
Used for culturing E. coli.
£294.68 - £432.48
Specifications
Content And Storage | Room Temperature |
---|---|
Format | Capsules |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12871670
|
Fisher BioReagents™
12871670 |
500 g |
£294.68
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
12861670
|
Fisher BioReagents™
12861670 |
25 x 20 g |
£432.48
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
- Pre-buffered
Specifications
Room Temperature | |
Capsules | |
Agar |