missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™ Syringe Filter, PES, 0.22 μm

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  Merck SLGP033NK

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10163282

  • £1800.00 / Pack of 1000
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility

Applications:
Buffer Filtration, Clarification of Aqueous Solutions

Specifications

Specifications

Single-use
0.22 μm
4.5 cm2
PP
<200 mL
33 mm
Millipore Express PLUS (PES)
Non-sterile
Analytical sample prep
1000Pack
33 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers