missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millex™ Syringe Filter, PES, 0.22 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
£452.00 - £1800.00
Specifications
Disposable | Single-use |
---|---|
Pore Size | 0.22 μm |
Sterility | Non-sterile |
For Use With (Application) | Analytical sample prep |
Housing Material | PP |
Product Code | Brand | Diameter (Metric) Filter | Quantity | Material | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Quantity | Material | Volume (Metric) | Price | Quantity & Availability | ||
11755223
|
Merck
SLGPX13NK |
13 mm | 1000Pack | Hydrophilic PES | <10 mL |
£1680.00
Pack of 1000 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||
11790453
|
Merck
SLGPX13NL |
13 mm | 100Pack | Hydrophilic PES | <10 mL |
£452.00
Pack of 100 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
10509952
|
Merck
SLGP033NB |
33 mm | 250Pack | Millipore Express PLUS (PES) | <200 mL |
£660.00
Pack of 250 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
10163282
|
Merck
SLGP033NK |
33 mm | 1000Pack | Millipore Express PLUS (PES) | <200 mL |
£1800.00
Pack of 1000 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications:
Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Single-use | |
Non-sterile | |
PP |
0.22 μm | |
Analytical sample prep |