Learn More
Merck Millex™ Syringe Filter, PES, 0.22 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Brand: Merck SLGPX13NL
287.13 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 71
Additional Details : Weight : 0.01000kg
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications:
Buffer Filtration, Clarification of Aqueous Solutions
Spécification
Green | |
Hydrophilic PES | |
Non-sterile | |
PP | |
male Luer slip | |
Syringe Filter | |
13 mm |
Single-use | |
0.22 μm | |
Analytical sample prep | |
male luer slip | |
100Pack | |
<10 mL | |
13 mm |