missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millicell Hanging Cell Culture Insert
Allows attachment or suspension cells to access media from both their apical and basolateral sides.
Brand: Merck MCSP24H48
This item is not returnable.
View return policy
Description
- Cell growth, structure, and function more closely mimic what occurs in vivo
- In addition, Millicell inserts make it possible to study both sides of the cell monolayer
- The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
- For co-culturing and permeability assays
- Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
- Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
- PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
- Individually blister packaged
Specifications
PET (Polyethylene Terephthalate) Filter Code | |
3 μm | |
50°C Max. Operating Temperature | |
6.5 mm | |
24-well Plates | |
Sterile |
Polystyrene | |
Cell Culture Insert | |
Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry | |
48/Pk | |
Clear | |
24 |