missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millicell Hanging Cell Culture Insert
Allows attachment or suspension cells to access media from both their apical and basolateral sides.
£282.00 - £444.00
Specifications
Description | PET (Polyethylene Terephthalate) Filter Code |
---|---|
Material | Polystyrene |
Type | Cell Culture Insert |
For Use With (Application) | Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry |
Quantity | 48/Pk |
Product Code | Brand | Pore Size | No. of Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. of Wells | Price | Quantity & Availability | ||||
15515960
|
Merck
MCHT06H48 |
0.4 μm | 6 | ||||||
15565960
|
Merck
MCRP06H48 |
1 μm | 6 | ||||||
15585960
|
Merck
MCSP06H48 |
3 μm | 6 | ||||||
15545960
|
Merck
MCMP06H48 |
5 μm | 6 | ||||||
15525960
|
Merck
MCHT12H48 |
0.4 μm | 12 | ||||||
15303328
|
Merck
MCRP12H48 |
1 μm | 12 | ||||||
15392839
|
Merck
MCSP12H48 |
3 μm | 12 | ||||||
15595950
|
Merck
MCEP12H48 |
8 μm | 12 | ||||||
15535960
|
Merck
MCHT24H48 |
0.4 μm | 24 | ||||||
15449779
|
Merck
MCRP24H48 |
1 μm | 24 | ||||||
15505970
|
Merck
MCSP24H48 |
3 μm | 24 | ||||||
15431055
|
Merck
MCMP24H48 |
5 μm | 24 | ||||||
15505960
|
Merck
MCEP24H48 |
8 μm | 24 | ||||||
Description
- Cell growth, structure, and function more closely mimic what occurs in vivo
- In addition, Millicell inserts make it possible to study both sides of the cell monolayer
- The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
- For co-culturing and permeability assays
- Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
- Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
- PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
- Individually blister packaged
Specifications
PET (Polyethylene Terephthalate) Filter Code | |
Cell Culture Insert | |
48/Pk | |
Sterile |
Polystyrene | |
Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry | |
Clear |